missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,3-dichlorobenzene, 98%

Catalog No. p-7026099
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2190414 25 g
AAB2190422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2190414 Supplier Thermo Scientific Chemicals Supplier No. B2190414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56961-77-4 | C6H3BrCl2 | 225.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56961-77-4
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.89
MDL Number MFCD00000536
InChI Key HVKCZUVMQPUWSX-UHFFFAOYSA-N
Synonym 2,3-dichlorobromobenzene, benzene, 1-bromo-2,3-dichloro, benzene, bromodichloro, 2,3-dichloro-1-bromobenzene, 1-bromo-2,3-dichloro-benzene, attercop-chm at113305, bromodichlorobenzene, pubchem3607, monobromodichlorobenzene, acmc-209lv2
PubChem CID 42066
IUPAC Name 1-bromo-2,3-dichlorobenzene
SMILES ClC1=CC=CC(Br)=C1Cl

Specifications

Melting Point 56°C to 60°C
Boiling Point 243°C
Quantity 25 g
Beilstein 2246806
Formula Weight 225.91
Percent Purity 98%
Chemical Name or Material 1-Bromo-2,3-dichlorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 260-476-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only