missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-1-propene (cis- and trans- mixture) 97.0+%, TCI America™

Supplier:  TCI America B103825G

 View more versions of this product

Catalog No. B103825G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Bromo-1-propene (cis- and trans- mixture)
-116°C
63°C
MFCD00000186,MFCD00082571
1993
NNQDMQVWOWCVEM-NSCUHMNNSA-N
(1E)-1-bromoprop-1-ene
643834
≥97.0% (GC)
590-14-7
Yellow
C3H5Br
25 g
cis-1-bromo-1-propene, z-1-bromo-1-propene, 1z-1-bromoprop-1-ene, 1-bromo-1-propene, cis-1-bromopropene, 1-propene, 1-bromo-, z, z-1-bromopropene, cis-propenyl bromide, inchi=1/c3h5br/c1-2-3-4/h2-3h,1h3/b3-2
C\C=C\Br
120.98
120.98
Liquid

Chemical Identifiers

590-14-7
120.98
NNQDMQVWOWCVEM-NSCUHMNNSA-N
643834
C\C=C\Br
C3H5Br
MFCD00000186,MFCD00082571
cis-1-bromo-1-propene, z-1-bromo-1-propene, 1z-1-bromoprop-1-ene, 1-bromo-1-propene, cis-1-bromopropene, 1-propene, 1-bromo-, z, z-1-bromopropene, cis-propenyl bromide, inchi=1/c3h5br/c1-2-3-4/h2-3h,1h3/b3-2
(1E)-1-bromoprop-1-ene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UC7080000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions