missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-1-propene (cis- and trans- mixture) 97.0+%, TCI America™

Catalog No. p-7118819
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10385G 5 g
B103825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B10385G Supplier TCI America Supplier No. B10385G

Chemical Identifiers

CAS 590-14-7
Molecular Formula C3H5Br
Molecular Weight (g/mol) 120.98
MDL Number MFCD00000186,MFCD00082571
InChI Key NNQDMQVWOWCVEM-NSCUHMNNSA-N
Synonym cis-1-bromo-1-propene, z-1-bromo-1-propene, 1z-1-bromoprop-1-ene, 1-bromo-1-propene, cis-1-bromopropene, 1-propene, 1-bromo-, z, z-1-bromopropene, cis-propenyl bromide, inchi=1/c3h5br/c1-2-3-4/h2-3h,1h3/b3-2
PubChem CID 643834
IUPAC Name (1E)-1-bromoprop-1-ene
SMILES C\C=C\Br

Specifications

Melting Point -116°C
Color Yellow
Boiling Point 63°C
Quantity 5 g
UN Number 1993
Formula Weight 120.98
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-1-propene (cis- and trans- mixture)
RTECSNumber UC7080000
TSCA Yes
Recommended Storage Refrigerator