missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-pyrrole-2-boronic acid pinacol ester, 97%

Catalog No. AAH6417106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6417106 5 g
AAH6417103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6417106 Supplier Thermo Scientific Chemicals Supplier No. H6417106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1072944-98-9 | C15H24BNO4 | 293.17 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1072944-98-9
Molecular Formula C15H24BNO4
Molecular Weight (g/mol) 293.17
MDL Number MFCD11504960
InChI Key DZQURIUJAGTAFW-UHFFFAOYSA-N
Synonym tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, 1-boc-pyrrole-2-boronic acid, pinacol ester, 1-boc-pyrrole-2-boronic acid pinacol ester, t-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole-1-carboxylate, pubchem23523, 1-boc-pyrrole-2-boronic acid,pinacol ester, 1-tert-butoxycarbonyl-1h-pyrrol-2-yl boronic acid pinacol ester, 1-boc-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole, tert-butyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate
PubChem CID 46739108
IUPAC Name tert-butyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole-1-carboxylate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CN2C(=O)OC(C)(C)C

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 293.17
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-Boc-pyrrole-2-boronic acid pinacol ester
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only