accessibility menu, dialog, popup

UserName

CAS RN 1072944-98-9

IUPAC Name: tert-butyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole-1-carboxylate
Synonyms: tert-butyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole-1-carboxylate
Molecular Weight (g/mol): 293.17
Molecular Formula: C15H24BNO4
InChi Key: DZQURIUJAGTAFW-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C=CC=C1B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results