Learn More
1-BOC-Piperidine, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 75844-69-8 |
|---|---|
| Molecular Formula | C10H19NO2 |
| Molecular Weight (g/mol) | 185.27 |
| InChI Key | RQCNHUCCQJMSRG-UHFFFAOYSA-N |
| Synonym | 1-boc-piperidine, n-boc-piperidine, 1-tert-butoxycarbonyl piperidine, 1-piperidinecarboxylic acid, 1,1-dimethylethyl ester, piperidine-1-carboxylic acid tert-butyl ester, tert-butyl piperidinecarboxylate, boc-piperidine, n-boc piperidine, pubchem7258, n-boc tetrahydropyridine |
| PubChem CID | 7010304 |
| IUPAC Name | tert-butyl piperidine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CCCCC1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC438080050
|
Thermo Scientific Chemicals
438080050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC438080250
|
Thermo Scientific Chemicals
438080250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 75844-69-8 | |
| 185.27 | |
| 1-boc-piperidine, n-boc-piperidine, 1-tert-butoxycarbonyl piperidine, 1-piperidinecarboxylic acid, 1,1-dimethylethyl ester, piperidine-1-carboxylic acid tert-butyl ester, tert-butyl piperidinecarboxylate, boc-piperidine, n-boc piperidine, pubchem7258, n-boc tetrahydropyridine | |
| tert-butyl piperidine-1-carboxylate |
| C10H19NO2 | |
| RQCNHUCCQJMSRG-UHFFFAOYSA-N | |
| 7010304 | |
| CC(C)(C)OC(=O)N1CCCCC1 |
Specifications
| 75844-69-8 | |
| 100.0 | |
| 86°C | |
| 96% min. (GC) | |
| C10H19NO2 | |
| 1-boc-piperidine, n-boc-piperidine, 1-tert-butoxycarbonyl piperidine, 1-piperidinecarboxylic acid, 1,1-dimethylethyl ester, piperidine-1-carboxylic acid tert-butyl ester, tert-butyl piperidinecarboxylate, boc-piperidine, n-boc piperidine, pubchem7258, n-boc tetrahydropyridine | |
| CC(C)(C)OC(=O)N1CCCCC1 | |
| 185.27 | |
| 185.27 | |
| 1-BOC-Piperidine |
| 96.0 | |
| 58°C (0.3 mbar) | |
| Authentic | |
| Glass bottle | |
| 5g | |
| RQCNHUCCQJMSRG-UHFFFAOYSA-N | |
| tert-butyl piperidine-1-carboxylate | |
| 7010304 | |
| 97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO â Research Use Only