missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-Piperidine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 438080250

 View more versions of this product

Catalog No. AC438080250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1-BOC-Piperidine
96.0
58°C (0.3 mbar)
Authentic
Glass bottle
25g
RQCNHUCCQJMSRG-UHFFFAOYSA-N
tert-butyl piperidine-1-carboxylate
7010304
97%
75844-69-8
100.0
86°C
96% min. (GC)
C10H19NO2
1-boc-piperidine, n-boc-piperidine, 1-tert-butoxycarbonyl piperidine, 1-piperidinecarboxylic acid, 1,1-dimethylethyl ester, piperidine-1-carboxylic acid tert-butyl ester, tert-butyl piperidinecarboxylate, boc-piperidine, n-boc piperidine, pubchem7258, n-boc tetrahydropyridine
CC(C)(C)OC(=O)N1CCCCC1
185.27
185.27

Chemical Identifiers

75844-69-8
185.27
1-boc-piperidine, n-boc-piperidine, 1-tert-butoxycarbonyl piperidine, 1-piperidinecarboxylic acid, 1,1-dimethylethyl ester, piperidine-1-carboxylic acid tert-butyl ester, tert-butyl piperidinecarboxylate, boc-piperidine, n-boc piperidine, pubchem7258, n-boc tetrahydropyridine
tert-butyl piperidine-1-carboxylate
C10H19NO2
RQCNHUCCQJMSRG-UHFFFAOYSA-N
7010304
CC(C)(C)OC(=O)N1CCCCC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only