missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-piperidine-4-carboxaldehyde, 97%

Catalog No. p-7035106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5281303 1 g
AAH5281306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5281303 Supplier Thermo Scientific Chemicals Supplier No. H5281303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 137076-22-3 | C11H19NO3 | 213.28 g/mol

1-Boc-piperidine-4-carboxaldehyde is used as reactant for synthesis of Pim-1 inhibitors, selective GPR119 agonists for type II diabetes, M-tropic (R5) HIV-1 replication inhibitors, HDAC inhibitors and selective 5-HT6 antagonists. It is also used as reactant for three-component vinylogous Mannich reactions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-piperidine-4-carboxaldehyde is used as reactant for synthesis of Pim-1 inhibitors, selective GPR119 agonists for type II diabetes, M-tropic (R5) HIV-1 replication inhibitors, HDAC inhibitors and selective 5-HT6 antagonists. It is also used as reactant for three-component vinylogous Mannich reactions

Solubility
Soluble in methanol.

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 137076-22-3
Molecular Formula C11H19NO3
Molecular Weight (g/mol) 213.28
MDL Number MFCD02179019
InChI Key JYUQEWCJWDGCRX-UHFFFAOYSA-N
Synonym 1-boc-4-piperidinecarboxaldehyde, 1-boc-4-formylpiperidine, 1-tert-butoxycarbonyl-4-piperidinecarboxaldehyde, 1-boc-piperidine-4-carboxaldehyde, n-boc-4-formylpiperidine, n-boc-4-piperidinecarboxaldehyde, 1-tert-butoxycarbonyl-4-formylpiperidine, 4-formyl-piperidine-1-carboxylic acid tert-butyl ester, n-boc-piperidine-4-carbaldehyde, 4-formylpiperidine-1-carboxylic acid tert-butyl ester
PubChem CID 1514430
IUPAC Name tert-butyl 4-formylpiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C=O

Specifications

Quantity 1 g
Sensitivity Air sensitive
Solubility Information Soluble in methanol.
Formula Weight 213.28
Percent Purity 97%
Chemical Name or Material 1-Boc-piperidine-4-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only