missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-N-Fmoc-D-tryptophan, 98%

CAS: 163619-04-3 | C31H30N2O6 | 526.59 g/mol

Supplier:  Thermo Scientific Chemicals H6218506

 View more versions of this product

Catalog No. AAH6218506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Boc-N-Fmoc-D-tryptophan
C31H30N2O6
5 g
fmoc-d-trp boc-oh, fmoc-d-trp boc-oh hplc, a-fmoc-n in-boc-d-tryptophan, fmoc-d-trp boc, fmoc-d-trp boc, pubchem10051, 1-boc-fmoc-d-tryptophan, 1-boc-fmoc-d-tryptophan, 2r-3-1-tert-butoxycarbonylindol-3-yl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, nalpha-fmoc-n in-boc-d-tryptophan, nalpha-fmoc-n in-boc-d-tryptophan
CC(C)(C)OC(=O)N1C=C(C[C@@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12
526.59
526.59
163619-04-3
MFCD00153367
7062970
ADOHASQZJSJZBT-AREMUKBSSA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-3-yl]propanoic acid
16213150
98%

Chemical Identifiers

163619-04-3
526.59
ADOHASQZJSJZBT-AREMUKBSSA-N
16213150
CC(C)(C)OC(=O)N1C=C(C[C@@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12
C31H30N2O6
MFCD00153367
fmoc-d-trp boc-oh, fmoc-d-trp boc-oh hplc, a-fmoc-n in-boc-d-tryptophan, fmoc-d-trp boc, fmoc-d-trp boc, pubchem10051, 1-boc-fmoc-d-tryptophan, 1-boc-fmoc-d-tryptophan, 2r-3-1-tert-butoxycarbonylindol-3-yl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, nalpha-fmoc-n in-boc-d-tryptophan, nalpha-fmoc-n in-boc-d-tryptophan
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-3-yl]propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only