missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-N-Fmoc-D-tryptophan, 98%

Catalog No. AAH6218503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6218503 1 g
AAH6218506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6218503 Supplier Thermo Scientific Chemicals Supplier No. H6218503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 163619-04-3 | C31H30N2O6 | 526.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 163619-04-3
Molecular Formula C31H30N2O6
Molecular Weight (g/mol) 526.59
MDL Number MFCD00153367
InChI Key ADOHASQZJSJZBT-AREMUKBSSA-N
Synonym fmoc-d-trp boc-oh, n, a-fmoc-n in-boc-d-tryptophan, fmoc-d-trp boc, ambotzfaa1339, pubchem10051, 1-boc-fmoc-d-tryptophan, 1-boc-n-fmoc-d-tryptophan, 2r-3-1-tert-butoxycarbonylindol-3-yl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, nalpha-fmoc-n in-boc-d-tryptophan, fmoc-d-trp boc-oh hplc
PubChem CID 16213150
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-3-yl]propanoic acid
SMILES CC(C)(C)OC(=O)N1C=C(C[C@@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12

Specifications

Quantity 1 g
Beilstein 7062970
Formula Weight 526.59
Percent Purity 98%
Chemical Name or Material 1-Boc-N-Fmoc-D-tryptophan
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only