Learn More
1-Boc-indoline-5-boronic acid pinacol ester, 97%
CAS: 837392-67-3 | C19H28BNO4 | 345.246 g/mol
$180.05 - $510.18
Chemical Identifiers
| CAS | 837392-67-3 |
|---|---|
| Molecular Formula | C19H28BNO4 |
| Molecular Weight (g/mol) | 345.246 |
| MDL Number | MFCD12408237 |
| InChI Key | OOORQXGLIKPNDK-UHFFFAOYSA-N |
| Synonym | 1-boc-indoline-5-boronic acid pinacol ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indoline-1-carboxylate, 1,1-dimethylethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-indole-1-carboxylate, 1h-indole-1-carboxylic acid, 2,3-dihydro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, 1,1-dimethylethyl ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-indole-1-carboxylate, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydroindole-1-carboxylate, 1-n-boc-5-bpin-indoline, amtb776, 1-tert-butoxycarbonyl-5-indolineboronic acid pinacol ester, 1-tert-butoxycarbonyl indolin-5-yl boronic acid pinacol ester |
| PubChem CID | 18451469 |
| IUPAC Name | tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroindole-1-carboxylate |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(CC3)C(=O)OC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH59436MD
|
Thermo Scientific Chemicals
H59436MD |
250 mg |
N/A
|
|
|||||
|
AAH5943603
|
Thermo Scientific Chemicals
H5943603 |
1 g |
N/A
|
|
|||||
Description
1-Boc-indoline-5-boronic acid pinacol ester is used as disinfector, tanning agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Boc-indoline-5-boronic acid pinacol ester is used as disinfector, tanning agent.
Solubility
Soluble in dichloromethane, ethyl acetate and methanol.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 837392-67-3 | |
| 345.246 | |
| OOORQXGLIKPNDK-UHFFFAOYSA-N | |
| 18451469 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(CC3)C(=O)OC(C)(C)C |
| C19H28BNO4 | |
| MFCD12408237 | |
| 1-boc-indoline-5-boronic acid pinacol ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indoline-1-carboxylate, 1,1-dimethylethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-indole-1-carboxylate, 1h-indole-1-carboxylic acid, 2,3-dihydro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, 1,1-dimethylethyl ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-indole-1-carboxylate, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydroindole-1-carboxylate, 1-n-boc-5-bpin-indoline, amtb776, 1-tert-butoxycarbonyl-5-indolineboronic acid pinacol ester, 1-tert-butoxycarbonyl indolin-5-yl boronic acid pinacol ester | |
| tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroindole-1-carboxylate |
Specifications
| 837392-67-3 | |
| MFCD12408237 | |
| 1-boc-indoline-5-boronic acid pinacol ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indoline-1-carboxylate, 1,1-dimethylethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-indole-1-carboxylate, 1h-indole-1-carboxylic acid, 2,3-dihydro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, 1,1-dimethylethyl ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-indole-1-carboxylate, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydroindole-1-carboxylate, 1-n-boc-5-bpin-indoline, amtb776, 1-tert-butoxycarbonyl-5-indolineboronic acid pinacol ester, 1-tert-butoxycarbonyl indolin-5-yl boronic acid pinacol ester | |
| OOORQXGLIKPNDK-UHFFFAOYSA-N | |
| tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroindole-1-carboxylate | |
| 18451469 | |
| 97% |
| C19H28BNO4 | |
| 250 mg | |
| Soluble in dichloromethane,ethyl acetate and methanol. | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(CC3)C(=O)OC(C)(C)C | |
| 345.246 | |
| 345.25 | |
| 1-Boc-indoline-5-boronic acid pinacol ester |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only