missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-indoline-5-boronic acid pinacol ester, 97%

CAS: 837392-67-3 | C19H28BNO4 | 345.246 g/mol

Supplier:  Thermo Scientific Chemicals H59436MD

 View more versions of this product

Catalog No. AAH59436MD


Only null left
Explore available promotions

Description

Description

1-Boc-indoline-5-boronic acid pinacol ester is used as disinfector, tanning agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-indoline-5-boronic acid pinacol ester is used as disinfector, tanning agent.

Solubility
Soluble in dichloromethane, ethyl acetate and methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

1-Boc-indoline-5-boronic acid pinacol ester
C19H28BNO4
250 mg
Soluble in dichloromethane,ethyl acetate and methanol.
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(CC3)C(=O)OC(C)(C)C
345.246
345.25
837392-67-3
MFCD12408237
1-boc-indoline-5-boronic acid pinacol ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indoline-1-carboxylate, 1,1-dimethylethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-indole-1-carboxylate, 1h-indole-1-carboxylic acid, 2,3-dihydro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, 1,1-dimethylethyl ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-indole-1-carboxylate, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydroindole-1-carboxylate, 1-n-boc-5-bpin-indoline, amtb776, 1-tert-butoxycarbonyl-5-indolineboronic acid pinacol ester, 1-tert-butoxycarbonyl indolin-5-yl boronic acid pinacol ester
OOORQXGLIKPNDK-UHFFFAOYSA-N
tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroindole-1-carboxylate
18451469
97%

Chemical Identifiers

837392-67-3
345.246
OOORQXGLIKPNDK-UHFFFAOYSA-N
18451469
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(CC3)C(=O)OC(C)(C)C
C19H28BNO4
MFCD12408237
1-boc-indoline-5-boronic acid pinacol ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indoline-1-carboxylate, 1,1-dimethylethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-indole-1-carboxylate, 1h-indole-1-carboxylic acid, 2,3-dihydro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, 1,1-dimethylethyl ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-indole-1-carboxylate, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydroindole-1-carboxylate, 1-n-boc-5-bpin-indoline, amtb776, 1-tert-butoxycarbonyl-5-indolineboronic acid pinacol ester, 1-tert-butoxycarbonyl indolin-5-yl boronic acid pinacol ester
tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroindole-1-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only