missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-indole, 97%

Catalog No. AAL1760106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1760106 5 g
AAL1760103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1760106 Supplier Thermo Scientific Chemicals Supplier No. L1760106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 75400-67-8 | C13H15NO2 | 217.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75400-67-8
Molecular Formula C13H15NO2
Molecular Weight (g/mol) 217.27
MDL Number MFCD02093939
InChI Key OWPIFQXNMLDXKW-UHFFFAOYSA-N
Synonym 1-boc-indole, tert-butyl 1h-indole-1-carboxylate, n-boc-indole, tert-butyl 1-indolecarboxylate, 1h-indole-1-carboxylic acid, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl indole, n-boc indole, pubchem7228, acmc-1bjgd, 1-tert-butoxycarbonylindole
PubChem CID 3532980
SMILES CC(C)(C)OC(=O)N1C=CC2=CC=CC=C12

Specifications

Density 1.07
Boiling Point 113°C to 114°C (0.2 mmHg)
Refractive Index 1.543
Quantity 5 g
Beilstein 4674239
IUPAC Name tert-butyl indole-1-carboxylate
Formula Weight 217.27
Percent Purity 97%
Chemical Name or Material 1-Boc-indole
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only