missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-homopiperazine, 98%

CAS: 112275-50-0 | C10H20N2O2 | 200.282 g/mol

Supplier:  Thermo Scientific Chemicals L1772806

 View more versions of this product

Catalog No. AAL1772806


Only null left

Description

Description

1-Boc-homopiperazine is used in synthesis of potent anticoagulant. It is also used to produce 4-(7,8,9,10-tetrahydro-6H-cyclohepta[b]quinolin-11-yl)-[1,4]diazepane-1-carboxylic acid tert-butyl ester by reaction with 11-bromo-7,8,9,10-tetrahydro-6H-cyclohepta[b]quinoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-homopiperazine is used in synthesis of potent anticoagulant. It is also used to produce 4-(7,8,9,10-tetrahydro-6H-cyclohepta[b]quinolin-11-yl)-[1,4]diazepane-1-carboxylic acid tert-butyl ester by reaction with 11-bromo-7,8,9,10-tetrahydro-6H-cyclohepta[b]quinoline.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. It is sensitive to air.
Specifications

Specifications

1-Boc-homopiperazine
90°C (0.5mmHg)
1.471
UN2735
1-boc-homopiperazine, 1-boc-hexahydro-1,4-diazepine, 1,4 diazepane-1-carboxylic acid tert-butyl ester, 1-tert-butoxycarbonyl homopiperazine, 1-boc-1,4-diazepane, n-boc-homopiperazine, boc-hopiz, boc-homopiperazine, n-tert-butoxycarbonyl homopiperazine, tert-butyl 1-homopiperazinecarboxylate
WDPWEXWMQDRXAL-UHFFFAOYSA-N
CC(C)(C)OC(=O)N1CCCNCC1
200.282
>110°C (230°F)
200.28
Air Sensitive
112275-50-0
C10H20N2O2
MFCD00276987
7581789
Not miscible or difficult to mix in water.
1.016
tert-butyl 1,4-diazepane-1-carboxylate
5 g
2756058
98%

Chemical Identifiers

112275-50-0
200.282
WDPWEXWMQDRXAL-UHFFFAOYSA-N
2756058
CC(C)(C)OC(=O)N1CCCNCC1
C10H20N2O2
MFCD00276987
1-boc-homopiperazine, 1-boc-hexahydro-1,4-diazepine, 1,4 diazepane-1-carboxylic acid tert-butyl ester, 1-tert-butoxycarbonyl homopiperazine, 1-boc-1,4-diazepane, n-boc-homopiperazine, boc-hopiz, boc-homopiperazine, n-tert-butoxycarbonyl homopiperazine, tert-butyl 1-homopiperazinecarboxylate
tert-butyl 1,4-diazepane-1-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only