missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-homopiperazine, 98%

Catalog No. p-7047145
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1772803 1 g
AAL1772806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1772803 Supplier Thermo Scientific Chemicals Supplier No. L1772803

CAS: 112275-50-0 | C10H20N2O2 | 200.282 g/mol

1-Boc-homopiperazine is used in synthesis of potent anticoagulant. It is also used to produce 4-(7,8,9,10-tetrahydro-6H-cyclohepta[b]quinolin-11-yl)-[1,4]diazepane-1-carboxylic acid tert-butyl ester by reaction with 11-bromo-7,8,9,10-tetrahydro-6H-cyclohepta[b]quinoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-homopiperazine is used in synthesis of potent anticoagulant. It is also used to produce 4-(7,8,9,10-tetrahydro-6H-cyclohepta[b]quinolin-11-yl)-[1,4]diazepane-1-carboxylic acid tert-butyl ester by reaction with 11-bromo-7,8,9,10-tetrahydro-6H-cyclohepta[b]quinoline.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. It is sensitive to air.

Chemical Identifiers

CAS 112275-50-0
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD00276987
InChI Key WDPWEXWMQDRXAL-UHFFFAOYSA-N
Synonym 1-boc-homopiperazine, 1-boc-hexahydro-1,4-diazepine, 1,4 diazepane-1-carboxylic acid tert-butyl ester, 1-tert-butoxycarbonyl homopiperazine, 1-boc-1,4-diazepane, n-boc-homopiperazine, boc-hopiz, boc-homopiperazine, n-tert-butoxycarbonyl homopiperazine, tert-butyl 1-homopiperazinecarboxylate
PubChem CID 2756058
IUPAC Name tert-butyl 1,4-diazepane-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCNCC1

Specifications

Boiling Point 90°C (0.5mmHg)
Refractive Index 1.471
UN Number UN2735
Beilstein 7581789
Solubility Information Not miscible or difficult to mix in water.
Density 1.016
Quantity 1 g
Flash Point >110°C (230°F)
Formula Weight 200.28
Sensitivity Air Sensitive
Percent Purity 98%
Chemical Name or Material 1-Boc-homopiperazine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only