Learn More
1-Boc-D-nipecotic acid, 97%
CAS: 163438-09-3 | C11H18NO4 | 228.27 g/mol
$187.12 - $725.53
Chemical Identifiers
| CAS | 163438-09-3 |
|---|---|
| Molecular Formula | C11H18NO4 |
| Molecular Weight (g/mol) | 228.27 |
| MDL Number | MFCD02179173 |
| InChI Key | NXILIHONWRXHFA-MRVPVSSYSA-M |
| Synonym | r-boc-nipecotic acid, r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, r-n-boc-piperidine-3-carboxylic acid, boc-d-nip-oh, r-1-boc-piperidine-3-carboxylic acid, r-1-boc-nipecotic acid, d-1-boc-nipecotic acid, n-boc-r-nipecotic acid, r-boc-nip, r-boc-nip-oh |
| PubChem CID | 643516 |
| SMILES | CC(C)(C)OC(=O)N1CCC[C@H](C1)C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5241606
|
Thermo Scientific Chemicals
H5241606 |
5 g |
Each for $187.12
|
|
|||||
|
AAH5241614
|
Thermo Scientific Chemicals
H5241614 |
25 g |
Each for $725.53
|
|
|||||
Description
1-Boc-D-nipecotic acid is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Boc-D-nipecotic acid is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 163438-09-3 | |
| 228.27 | |
| NXILIHONWRXHFA-MRVPVSSYSA-M | |
| 643516 |
| C11H18NO4 | |
| MFCD02179173 | |
| r-boc-nipecotic acid, r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, r-n-boc-piperidine-3-carboxylic acid, boc-d-nip-oh, r-1-boc-piperidine-3-carboxylic acid, r-1-boc-nipecotic acid, d-1-boc-nipecotic acid, n-boc-r-nipecotic acid, r-boc-nip, r-boc-nip-oh | |
| CC(C)(C)OC(=O)N1CCC[C@H](C1)C([O-])=O |
Specifications
| 163438-09-3 | |
| MFCD02179173 | |
| 5539297 | |
| NXILIHONWRXHFA-MRVPVSSYSA-M | |
| (3R)-1-[(tert-butoxy)carbonyl]piperidine-3-carboxylate | |
| 643516 | |
| 97% |
| C11H18NO4 | |
| 5 g | |
| r-boc-nipecotic acid, r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, r-n-boc-piperidine-3-carboxylic acid, boc-d-nip-oh, r-1-boc-piperidine-3-carboxylic acid, r-1-boc-nipecotic acid, d-1-boc-nipecotic acid, n-boc-r-nipecotic acid, r-boc-nip, r-boc-nip-oh | |
| CC(C)(C)OC(=O)N1CCC[C@H](C1)C([O-])=O | |
| 228.27 | |
| 229.28 | |
| 1-Boc-D-nipecotic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only