CAS RN 163438-09-3
IUPAC Name:
(3R)-1-[(tert-butoxy)carbonyl]piperidine-3-carboxylate
Synonyms:
(3R)-1-(tert-butoxycarbonyl)piperidine-3-carboxylate
Molecular Weight (g/mol):
228.27
Molecular Formula:
C11H18NO4
InChi Key:
NXILIHONWRXHFA-MRVPVSSYSA-M
SMILES:
CC(C)(C)OC(=O)N1CCC[C@H](C1)C([O-])=O
1
–
1
of
1
results
1
Promotions Available
1-Boc-D-nipecotic acid, 97%
1-Boc-D-nipecotic acid, 97%
CAS: 163438-09-3 Molecular Formula: C11H18NO4 Molecular Weight (g/mol): 228.27 MDL Number: MFCD02179173 InChI Key: NXILIHONWRXHFA-MRVPVSSYSA-M Synonym: r-boc-nipecotic acid,r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid,r-n-boc-piperidine-3-carboxylic acid,boc-d-nip-oh,r-1-boc-piperidine-3-carboxylic acid,r-1-boc-nipecotic acid,d-1-boc-nipecotic acid,n-boc-r-nipecotic acid,r-boc-nip,r-boc-nip-oh PubChem CID: 643516 SMILES: CC(C)(C)OC(=O)N1CCC[C@H](C1)C([O-])=O