missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-azetidine-3-carboxylic acid, 97%

Catalog No. p-7030455
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2881703 1 g
AAH2881706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2881703 Supplier Thermo Scientific Chemicals Supplier No. H2881703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 142253-55-2 | C9H14NO4 | 200.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 142253-55-2
Molecular Formula C9H14NO4
Molecular Weight (g/mol) 200.22
MDL Number MFCD01860897
InChI Key NCADHSLPNSTDMJ-UHFFFAOYSA-M
Synonym 1-boc-azetidine-3-carboxylic acid, 1-n-boc-3-azetidinecarboxylic acid, n-boc-azetidine-3-carboxylic acid, boc-azetidine-3-carboxylic acid, 1-tert-butoxycarbonyl azetidine-3-carboxylic acid, 1-tert-butoxy carbonyl azetidine-3-carboxylic acid, 1-n-boc-azetidine-3-carboxylic acid, boc-aze 3-oh, azetidine-1,3-dicarboxylic acid mono-tert-butyl ester, n-boc-3-azetidine carboxylic acid
PubChem CID 2755981
SMILES CC(C)(C)OC(=O)N1CC(C1)C([O-])=O

Specifications

Melting Point 98°C to 105°C
IUPAC Name 1-[(tert-butoxy)carbonyl]azetidine-3-carboxylate
Quantity 1 g
Formula Weight 201.22
Percent Purity 97%
Chemical Name or Material 1-Boc-azetidine-3-carboxylic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA No

RUO – Research Use Only