Learn More
1-Boc-6-cyanoindole-2-boronic acid, 96%
CAS: 913835-67-3 | C14H15BN2O4 | 286.09 g/mol
Supplier: Thermo Scientific Chemicals H5252003
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-Boc-6-cyanoindole-2-boronic acid | |
| 219°C to 221°C | |
| MFCD08436056 | |
| 1-boc-6-cyanoindole-2-boronic acid, 1-tert-butoxycarbonyl-6-cyano-1h-indol-2-yl boronic acid, 1-tert-butoxycarbonyl-6-cyano-1h-indole-2-boronic acid, 6-cyano-1h-indole-2-boronic acid, n-boc protected, 1-tert-butoxycarbonyl-6-cyanoindol-2-ylboronic acid, acmc-209r9t, 6-cyano-1h-indol-2-ylboronic acid, n-boc protected, 1-tert-butoxycarbonyl-6-cyano-1h-indol-2-yl boronicacid, 1-tert-butoxycarbonyl-2-dihydroxyboryl-1h-indole-6-carbonitrile | |
| CC(C)(C)OC(=O)N1C(=CC2=CC=C(C=C12)C#N)B(O)O | |
| 286.09 | |
| 286.09 |
| 913835-67-3 | |
| C14H15BN2O4 | |
| 1 g | |
| CZEBTZMIQZVUSE-UHFFFAOYSA-N | |
| [6-cyano-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid | |
| 44119347 | |
| 96% |
Chemical Identifiers
| 913835-67-3 | |
| 286.09 | |
| CZEBTZMIQZVUSE-UHFFFAOYSA-N | |
| 44119347 | |
| CC(C)(C)OC(=O)N1C(=CC2=CC=C(C=C12)C#N)B(O)O |
| C14H15BN2O4 | |
| MFCD08436056 | |
| 1-boc-6-cyanoindole-2-boronic acid, 1-tert-butoxycarbonyl-6-cyano-1h-indol-2-yl boronic acid, 1-tert-butoxycarbonyl-6-cyano-1h-indole-2-boronic acid, 6-cyano-1h-indole-2-boronic acid, n-boc protected, 1-tert-butoxycarbonyl-6-cyanoindol-2-ylboronic acid, acmc-209r9t, 6-cyano-1h-indol-2-ylboronic acid, n-boc protected, 1-tert-butoxycarbonyl-6-cyano-1h-indol-2-yl boronicacid, 1-tert-butoxycarbonyl-2-dihydroxyboryl-1h-indole-6-carbonitrile | |
| [6-cyano-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only