missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-6-cyanoindole-2-boronic acid, 96%

Catalog No. p-7034821
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52520MD 250 mg
AAH5252003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52520MD Supplier Thermo Scientific Chemicals Supplier No. H52520MD
Only null left

CAS: 913835-67-3 | C14H15BN2O4 | 286.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 913835-67-3
Molecular Formula C14H15BN2O4
Molecular Weight (g/mol) 286.09
MDL Number MFCD08436056
InChI Key CZEBTZMIQZVUSE-UHFFFAOYSA-N
Synonym 1-boc-6-cyanoindole-2-boronic acid, 1-tert-butoxycarbonyl-6-cyano-1h-indol-2-yl boronic acid, 1-tert-butoxycarbonyl-6-cyano-1h-indole-2-boronic acid, 6-cyano-1h-indole-2-boronic acid, n-boc protected, 1-tert-butoxycarbonyl-6-cyanoindol-2-ylboronic acid, acmc-209r9t, 6-cyano-1h-indol-2-ylboronic acid, n-boc protected, 1-tert-butoxycarbonyl-6-cyano-1h-indol-2-yl boronicacid, 1-tert-butoxycarbonyl-2-dihydroxyboryl-1h-indole-6-carbonitrile
PubChem CID 44119347
IUPAC Name [6-cyano-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid
SMILES CC(C)(C)OC(=O)N1C(=CC2=CC=C(C=C12)C#N)B(O)O

Specifications

Melting Point 219°C to 221°C
Quantity 250 mg
Formula Weight 286.09
Percent Purity 96%
Chemical Name or Material 1-Boc-6-cyanoindole-2-boronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only