missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-5-bromoindoline, 97%

Catalog No. p-7039863
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59341MD 250 mg
AAH5934103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59341MD Supplier Thermo Scientific Chemicals Supplier No. H59341MD
Only null left

CAS: 261732-38-1 | C13H16BrNO2 | 298.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 261732-38-1
Molecular Formula C13H16BrNO2
Molecular Weight (g/mol) 298.18
MDL Number MFCD08059280
InChI Key UOCVSZYBRMGQOL-UHFFFAOYSA-N
Synonym tert-butyl 5-bromoindoline-1-carboxylate, 1-boc-5-bromoindoline, n-boc-5-bromoindoline, tert-butyl 5-bromo-2,3-dihydro-1h-indole-1-carboxylate, 5-bromo-2,3-dihydro-indole-1-carboxylic acid tert-butyl ester, 1h-indole-1-carboxylic acid, 5-bromo-2,3-dihydro-, 1,1-dimethylethyl ester, acmc-20aixt, amth084, 5-bromo-1-t-butyloxycarbonylindoline
PubChem CID 21865438
IUPAC Name tert-butyl 5-bromo-2,3-dihydroindole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC2=C1C=CC(=C2)Br

Specifications

Quantity 250 mg
Formula Weight 298.18
Percent Purity 97%
Chemical Name or Material 1-Boc-5-bromoindoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only