missing translation for 'onlineSavingsMsg'
Learn More

(1-Boc-4-piperidinyloxy)acetic acid, 95%, Thermo Scientific Chemicals

Catalog No. AAH3210203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3210203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3210203 Supplier Thermo Scientific Chemicals Supplier No. H3210203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 161948-70-5 | Molecular Formula: C12H21NO5 | Molecular Weight: 259.302 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 161948-70-5
Molecular Formula C12H21NO5
Molecular Weight (g/mol) 259.302
MDL Number MFCD02178987
InChI Key YHUAHIMRWSVXCN-UHFFFAOYSA-N
Synonym n-boc-4-carboxymethoxypiperidine, n-boc-4-carboxymethoxy-piperidine, 4-carboxymethoxy-piperidine-1-carboxylic acid tert-butyl ester, 1-boc-4-piperidinyloxy acetic acid, 2-1-tert-butoxy carbonyl piperidin-4-yl oxy acetic acid, 1-tert-butoxycarbonyl piperidin-4-yl oxy acetic acid, 1-boc-4-carboxymethoxy-piperidine, 2-1-tert-butoxycarbonyl piperidin-4-yloxy acetic acid, 2-1-tert-butoxycarbonyl piperidin-4-yloxy aceticacid, 2-1-tert-butoxycarbonyl-4-piperidyl oxy acetic acid
PubChem CID 1502082
IUPAC Name 2-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]oxyacetic acid
SMILES CC(C)(C)OC(=O)N1CCC(CC1)OCC(=O)O

Specifications

Melting Point 105°C to 107°C
Quantity 1 g
Formula Weight 259.3
Percent Purity 95%
Chemical Name or Material (1-Boc-4-piperidinyloxy)acetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only