missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(4-aminophenyl)piperazine, 97%

Catalog No. AAH6319103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6319103 1 g
AAH6319106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6319103 Supplier Thermo Scientific Chemicals Supplier No. H6319103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 170911-92-9 | C15H23N3O2 | 277.368 g/mol

1-Boc-4-(4-aminophenyl)piperazine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-4-(4-aminophenyl)piperazine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 170911-92-9
Molecular Formula C15H23N3O2
Molecular Weight (g/mol) 277.368
MDL Number MFCD04115065
InChI Key RXFHRKPNLPBDGE-UHFFFAOYSA-N
Synonym tert-butyl 4-4-aminophenyl piperazine-1-carboxylate, 4-4-aminophenyl piperazine-1-carboxylic acid tert-butyl ester, 4-4-boc-piperazin-1-yl aniline, 1-boc-4-4-aminophenyl piperazine, 1-piperazinecarboxylic acid, 4-4-aminophenyl-, 1,1-dimethylethyl ester, 4-4-aminophenyl-piperazine-1-carboxylic acid tert-butyl ester, 1-boc-4-4'-aminophenyl piperazine dihydrochloride, 1-boc-4-4-aminophenyl-piperazine, pubchem12198
PubChem CID 11011301
IUPAC Name tert-butyl 4-(4-aminophenyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)N

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 277.36
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-4-(4-aminophenyl)piperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only