accessibility menu, dialog, popup

UserName

CH3H3CCH3OONNNH2

CAS RN 170911-92-9

IUPAC Name: tert-butyl 4-(4-aminophenyl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(4-aminophenyl)piperazine-1-carboxylate
Molecular Weight (g/mol): 277.37
Molecular Formula: C15H23N3O2
InChi Key: RXFHRKPNLPBDGE-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results