missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Boc-3-methylpiperazine, 97%

Catalog No. AAH5280914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5280914 25 g
AAH5280906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5280914 Supplier Thermo Scientific Chemicals Supplier No. H5280914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 120737-59-9 | C10H20N2O2 | 200.282 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 120737-59-9
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD03001706
InChI Key FMLPQHJYUZTHQS-UHFFFAOYSA-N
Synonym 4-n-boc-2-methyl-piperazine, 1-boc-3-methylpiperazine, 4-n-boc-2-methylpiperazine, 4-boc-2-methyl-piperazine, 1-boc-3-methyl-piperazine, 3-methyl-piperazine-1-carboxylic acid tert-butyl ester, 4-boc-2-methylpiperazine, tert-butyl 3-methyl-1-piperazinecarboxylate, 2-methylpiperazine, n4-boc protected, 2-methyl-4-boc-piperazine
PubChem CID 2756810
IUPAC Name tert-butyl 3-methylpiperazine-1-carboxylate
SMILES CC1CN(CCN1)C(=O)OC(C)(C)C

Specifications

Quantity 25 g
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material (+/-)-1-Boc-3-methylpiperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only