CAS RN 120737-59-9
IUPAC Name:
tert-butyl 3-methylpiperazine-1-carboxylate
Synonyms:
tert-butyl 3-methylpiperazine-1-carboxylate
Molecular Weight (g/mol):
200.28
Molecular Formula:
C10H20N2O2
InChi Key:
FMLPQHJYUZTHQS-UHFFFAOYNA-N
SMILES:
CC1CN(CCN1)C(=O)OC(C)(C)C
1
–
1
of
1
results
1
(+/-)-1-Boc-3-methylpiperazine, 97%
CAS: 120737-59-9 Molecular Formula: C10H20N2O2 Molecular Weight (g/mol): 200.282 MDL Number: MFCD03001706 InChI Key: FMLPQHJYUZTHQS-UHFFFAOYSA-N Synonym: 4-n-boc-2-methyl-piperazine,1-boc-3-methylpiperazine,4-n-boc-2-methylpiperazine,4-boc-2-methyl-piperazine,1-boc-3-methyl-piperazine,3-methyl-piperazine-1-carboxylic acid tert-butyl ester,4-boc-2-methylpiperazine,tert-butyl 3-methyl-1-piperazinecarboxylate,2-methylpiperazine, n4-boc protected,2-methyl-4-boc-piperazine PubChem CID: 2756810 IUPAC Name: tert-butyl 3-methylpiperazine-1-carboxylate SMILES: CC1CN(CCN1)C(=O)OC(C)(C)C