missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-(methylamino)piperidine, 97%

Catalog No. AAH5277303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5277303 1 g
AAH5277306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5277303 Supplier Thermo Scientific Chemicals Supplier No. H5277303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 392331-89-4 | C11H22N2O2 | 214.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 392331-89-4
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.31
MDL Number MFCD04114980,MFCD09752963
InChI Key XRRRUOWSHGFPTI-UHFFFAOYNA-N
Synonym 1-boc-3-methylaminopiperidine, tert-butyl 3-methylamino piperidine-1-carboxylate, 1-boc-3-methylaminopieridine, n-boc-3-methylamino piperidine, 1-n-boc-3-methylaminopiperidine, 1-boc-3-methykaminopiperidine, n-boc-3-methylaminopiperidine, 3-methylamino-piperidine-1-carboxylic acid tert-butyl ester, 1-boc-3-methylamino piperidine, 3-methylamino piperidine-1-carboxylic acid tert-butyl ester
PubChem CID 17750458
IUPAC Name tert-butyl 3-(methylamino)piperidine-1-carboxylate
SMILES CNC1CCCN(C1)C(=O)OC(C)(C)C

Specifications

Boiling Point 95°C to 99°C (0.6 mmHg)
Quantity 1 g
Formula Weight 214.31
Percent Purity 97%
Chemical Name or Material 1-Boc-3-(methylamino)piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only