accessibility menu, dialog, popup

UserName

CH3HNNOOH3CH3CCH3

CAS RN 392331-89-4

IUPAC Name: tert-butyl 3-(methylamino)piperidine-1-carboxylate
Synonyms: tert-butyl 3-(methylamino)piperidine-1-carboxylate
Molecular Weight (g/mol): 214.31
Molecular Formula: C11H22N2O2
InChi Key: XRRRUOWSHGFPTI-UHFFFAOYNA-N
SMILES: CNC1CCCN(C1)C(=O)OC(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)