Learn More
1-Boc-3-(methylamino)azetidine, 95%, Thermo Scientific Chemicals
$311.63 - $311.63
Chemical Identifiers
| CAS | 454703-20-9 |
|---|---|
| Molecular Formula | C9H18N2O2 |
| Molecular Weight (g/mol) | 186.255 |
| MDL Number | MFCD08752496 |
| InChI Key | CHRBSEYIEDTNSC-UHFFFAOYSA-N |
| Synonym | tert-butyl 3-methylamino azetidine-1-carboxylate, 1-boc-3-methylamino azetidine, 1-n-boc-3-methylamino azetidine, 1-azetidinecarboxylic acid, 3-methylamino-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-methylamino azetidine, 1-boc-3-methylaminoazetidine, 1-azetidinecarboxylicacid, 3-methylamino-, 1,1-dimethylethyl ester, 1-boc-3-methylaminoazatidine, 1-boc-3-methyl amino azatidine hydrochloride, 3-methylamino-1-azetidinecarboxylic acid tert-butyl ester |
| PubChem CID | 23153172 |
| IUPAC Name | tert-butyl 3-(methylamino)azetidine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CC(C1)NC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5795103
|
Thermo Scientific Chemicals
H5795103 |
1 g |
Each for $311.63
|
|
|||||
Description
1-Boc-3-(methylamino)azetidine is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Boc-3-(methylamino)azetidine is used as pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with oxidizing agents.
Chemical Identifiers
| 454703-20-9 | |
| 186.255 | |
| CHRBSEYIEDTNSC-UHFFFAOYSA-N | |
| 23153172 | |
| CC(C)(C)OC(=O)N1CC(C1)NC |
| C9H18N2O2 | |
| MFCD08752496 | |
| tert-butyl 3-methylamino azetidine-1-carboxylate, 1-boc-3-methylamino azetidine, 1-n-boc-3-methylamino azetidine, 1-azetidinecarboxylic acid, 3-methylamino-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-methylamino azetidine, 1-boc-3-methylaminoazetidine, 1-azetidinecarboxylicacid, 3-methylamino-, 1,1-dimethylethyl ester, 1-boc-3-methylaminoazatidine, 1-boc-3-methyl amino azatidine hydrochloride, 3-methylamino-1-azetidinecarboxylic acid tert-butyl ester | |
| tert-butyl 3-(methylamino)azetidine-1-carboxylate |
Specifications
| 454703-20-9 | |
| MFCD08752496 | |
| Slightly soluble in water. | |
| CC(C)(C)OC(=O)N1CC(C1)NC | |
| 1 g | |
| 23153172 | |
| Air Sensitive | |
| 1-Boc-3-(methylamino)azetidine |
| C9H18N2O2 | |
| tert-butyl 3-methylamino azetidine-1-carboxylate, 1-boc-3-methylamino azetidine, 1-n-boc-3-methylamino azetidine, 1-azetidinecarboxylic acid, 3-methylamino-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-methylamino azetidine, 1-boc-3-methylaminoazetidine, 1-azetidinecarboxylicacid, 3-methylamino-, 1,1-dimethylethyl ester, 1-boc-3-methylaminoazatidine, 1-boc-3-methyl amino azatidine hydrochloride, 3-methylamino-1-azetidinecarboxylic acid tert-butyl ester | |
| CHRBSEYIEDTNSC-UHFFFAOYSA-N | |
| tert-butyl 3-(methylamino)azetidine-1-carboxylate | |
| 186.255 | |
| 186.25 | |
| 95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only