accessibility menu, dialog, popup

UserName

CH3H3CCH3OONHNCH3

CAS RN 454703-20-9

IUPAC Name: tert-butyl 3-(methylamino)azetidine-1-carboxylate
Synonyms: tert-butyl 3-(methylamino)azetidine-1-carboxylate
Molecular Weight (g/mol): 186.26
Molecular Formula: C9H18N2O2
InChi Key: CHRBSEYIEDTNSC-UHFFFAOYSA-N
SMILES: CNC1CN(C1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results