missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-hydroxyazetidine, 97%

CAS: 141699-55-0 | C8H15NO3 | 173.212 g/mol

$97.62 - $300.10

Chemical Identifiers

CAS 141699-55-0
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.212
MDL Number MFCD04115305
InChI Key XRRXRQJQQKMFBC-UHFFFAOYSA-N
Synonym 1-n-boc-3-hydroxyazetidine, n-boc-3-hydroxyazetidine, 1-boc-3-hydroxyazetidine, 1-boc-3-hydroxy azetidine, 1-boc-3-azetidinol, 1-tert-butoxycarbonyl-3-hydroxyazetidine, 3-hydroxy-azetidine-1-carboxylic acid tert-butyl ester, n-boc-3-hydroxy azetidine, n-boc-azetidin-3-ol, 3-hydroxyazetidine-1-carboxylic acid tert-butyl ester
PubChem CID 2756801
IUPAC Name tert-butyl 3-hydroxyazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2890203
View Documents
Thermo Scientific Chemicals
H2890203
1 g
Each for $97.62
Only null left
 
AAH2890206
View Documents
Thermo Scientific Chemicals
H2890206
5 g
Each for $300.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

141699-55-0
173.212
XRRXRQJQQKMFBC-UHFFFAOYSA-N
2756801
CC(C)(C)OC(=O)N1CC(C1)O
C8H15NO3
MFCD04115305
1-n-boc-3-hydroxyazetidine, n-boc-3-hydroxyazetidine, 1-boc-3-hydroxyazetidine, 1-boc-3-hydroxy azetidine, 1-boc-3-azetidinol, 1-tert-butoxycarbonyl-3-hydroxyazetidine, 3-hydroxy-azetidine-1-carboxylic acid tert-butyl ester, n-boc-3-hydroxy azetidine, n-boc-azetidin-3-ol, 3-hydroxyazetidine-1-carboxylic acid tert-butyl ester
tert-butyl 3-hydroxyazetidine-1-carboxylate

Specifications

40°C to 47°C
C8H15NO3
1-n-boc-3-hydroxyazetidine, n-boc-3-hydroxyazetidine, 1-boc-3-hydroxyazetidine, 1-boc-3-hydroxy azetidine, 1-boc-3-azetidinol, 1-tert-butoxycarbonyl-3-hydroxyazetidine, 3-hydroxy-azetidine-1-carboxylic acid tert-butyl ester, n-boc-3-hydroxy azetidine, n-boc-azetidin-3-ol, 3-hydroxyazetidine-1-carboxylic acid tert-butyl ester
CC(C)(C)OC(=O)N1CC(C1)O
1 g
2756801
97%
141699-55-0
MFCD04115305
XRRXRQJQQKMFBC-UHFFFAOYSA-N
tert-butyl 3-hydroxyazetidine-1-carboxylate
173.212
173.21
1-Boc-3-hydroxyazetidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only