missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-hydroxyazetidine, 97%

CAS: 141699-55-0 | C8H15NO3 | 173.212 g/mol

Supplier:  Thermo Scientific Chemicals H2890203

 View more versions of this product

Catalog No. AAH2890203


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

40°C to 47°C
141699-55-0
MFCD04115305
XRRXRQJQQKMFBC-UHFFFAOYSA-N
tert-butyl 3-hydroxyazetidine-1-carboxylate
173.212
173.21
1-Boc-3-hydroxyazetidine
C8H15NO3
1-n-boc-3-hydroxyazetidine, n-boc-3-hydroxyazetidine, 1-boc-3-hydroxyazetidine, 1-boc-3-hydroxy azetidine, 1-boc-3-azetidinol, 1-tert-butoxycarbonyl-3-hydroxyazetidine, 3-hydroxy-azetidine-1-carboxylic acid tert-butyl ester, n-boc-3-hydroxy azetidine, n-boc-azetidin-3-ol, 3-hydroxyazetidine-1-carboxylic acid tert-butyl ester
CC(C)(C)OC(=O)N1CC(C1)O
1 g
2756801
97%

Chemical Identifiers

141699-55-0
173.212
XRRXRQJQQKMFBC-UHFFFAOYSA-N
2756801
CC(C)(C)OC(=O)N1CC(C1)O
C8H15NO3
MFCD04115305
1-n-boc-3-hydroxyazetidine, n-boc-3-hydroxyazetidine, 1-boc-3-hydroxyazetidine, 1-boc-3-hydroxy azetidine, 1-boc-3-azetidinol, 1-tert-butoxycarbonyl-3-hydroxyazetidine, 3-hydroxy-azetidine-1-carboxylic acid tert-butyl ester, n-boc-3-hydroxy azetidine, n-boc-azetidin-3-ol, 3-hydroxyazetidine-1-carboxylic acid tert-butyl ester
tert-butyl 3-hydroxyazetidine-1-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only