missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-fluoro-4-pyrrolidinone

CAS: 845894-03-3 | C9H14FNO3 | 203.21 g/mol

$347.77 - $983.27

Chemical Identifiers

CAS 845894-03-3
Molecular Formula C9H14FNO3
Molecular Weight (g/mol) 203.21
MDL Number MFCD08274459
InChI Key HPHVNLXMQXEDFX-UHFFFAOYNA-N
Synonym 1-boc-3-fluoro-4-pyrrolidinone, boc-3-fluoro-4-oxopyrrolidine, 3-fluoro-4-oxopyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-4-fluoro-3-pyrrolidinone, n-t-boc-3-fluoro-4-oxopyrrolidine, tert-butyl3-fluoro-4-oxopyrrolidine-1-carboxylate
PubChem CID 45158916
IUPAC Name tert-butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(F)C(=O)C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH27082MD
View Documents
Thermo Scientific Chemicals
H27082MD
250 mg N/A
Only null left
 
AAH2708203
View Documents
Thermo Scientific Chemicals
H2708203
1 g N/A
Only null left
 
Description

Description

1-Boc-3-fluoro-4-pyrrolidinone is used as an intermediate in the manufacture of various substances such as pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-3-fluoro-4-pyrrolidinone is used as an intermediate in the manufacture of various substances such as pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications

Chemical Identifiers

845894-03-3
203.21
HPHVNLXMQXEDFX-UHFFFAOYNA-N
45158916
CC(C)(C)OC(=O)N1CC(F)C(=O)C1
C9H14FNO3
MFCD08274459
1-boc-3-fluoro-4-pyrrolidinone, boc-3-fluoro-4-oxopyrrolidine, 3-fluoro-4-oxopyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-4-fluoro-3-pyrrolidinone, n-t-boc-3-fluoro-4-oxopyrrolidine, tert-butyl3-fluoro-4-oxopyrrolidine-1-carboxylate
tert-butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate

Specifications

845894-03-3
MFCD08274459
1-boc-3-fluoro-4-pyrrolidinone, boc-3-fluoro-4-oxopyrrolidine, 3-fluoro-4-oxopyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-4-fluoro-3-pyrrolidinone, n-t-boc-3-fluoro-4-oxopyrrolidine, tert-butyl3-fluoro-4-oxopyrrolidine-1-carboxylate
HPHVNLXMQXEDFX-UHFFFAOYNA-N
tert-butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate
45158916
1-Boc-3-fluoro-4-pyrrolidinone
C9H14FNO3
250 mg
Slightly soluble in water.
CC(C)(C)OC(=O)N1CC(F)C(=O)C1
203.21
203.22
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only