Learn More
1-Boc-3-fluoro-4-pyrrolidinone
CAS: 845894-03-3 | C9H14FNO3 | 203.21 g/mol
Supplier: Thermo Scientific Chemicals H2708203
Description
1-Boc-3-fluoro-4-pyrrolidinone is used as an intermediate in the manufacture of various substances such as pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Boc-3-fluoro-4-pyrrolidinone is used as an intermediate in the manufacture of various substances such as pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications
| 1-Boc-3-fluoro-4-pyrrolidinone | |
| C9H14FNO3 | |
| 1 g | |
| Slightly soluble in water. | |
| CC(C)(C)OC(=O)N1CC(F)C(=O)C1 | |
| 203.21 | |
| 203.22 |
| 845894-03-3 | |
| MFCD08274459 | |
| 1-boc-3-fluoro-4-pyrrolidinone, boc-3-fluoro-4-oxopyrrolidine, 3-fluoro-4-oxopyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-4-fluoro-3-pyrrolidinone, n-t-boc-3-fluoro-4-oxopyrrolidine, tert-butyl3-fluoro-4-oxopyrrolidine-1-carboxylate | |
| HPHVNLXMQXEDFX-UHFFFAOYNA-N | |
| tert-butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate | |
| 45158916 |
Chemical Identifiers
| 845894-03-3 | |
| 203.21 | |
| HPHVNLXMQXEDFX-UHFFFAOYNA-N | |
| 45158916 | |
| CC(C)(C)OC(=O)N1CC(F)C(=O)C1 |
| C9H14FNO3 | |
| MFCD08274459 | |
| 1-boc-3-fluoro-4-pyrrolidinone, boc-3-fluoro-4-oxopyrrolidine, 3-fluoro-4-oxopyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-4-fluoro-3-pyrrolidinone, n-t-boc-3-fluoro-4-oxopyrrolidine, tert-butyl3-fluoro-4-oxopyrrolidine-1-carboxylate | |
| tert-butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate |
Safety and Handling
TSCA : No
RUO – Research Use Only