missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-3-(bromomethyl)azetidine, 95%

Catalog No. AC452160050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452160050 5 g
AC452160010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC452160050 Supplier Thermo Scientific Chemicals Supplier No. 452160050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 253176-93-1 | C9H16BrNO2 | 250.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 253176-93-1
Molecular Formula C9H16BrNO2
Molecular Weight (g/mol) 250.14
MDL Number MFCD16556174
InChI Key PUKCUGDJEPVLPR-UHFFFAOYSA-N
Synonym 1-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl azetidine-1-carboxylate, 1-boc-3-bromomethylazetidine, n-tert-butoxycarbonyl-3-bromomethylazetidine, 3-bromomethyl-azetidine-1-carboxylic acid tert-butyl ester, acmc-209d8t, 1-n-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl-1-azetidinecarboxylate, tert-butyl 3-bromomethyl ;azetidine-1-carboxylate, 2-methyl-2-propanyl 3-bromomethyl-1-azetidinecarboxylate
PubChem CID 53350331
IUPAC Name tert-butyl 3-(bromomethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(CBr)C1

Specifications

Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 250.13
Percent Purity 95%
Chemical Name or Material 1-BOC-3-(bromomethyl)azetidine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

RUO – Research Use Only