Learn More
1-Boc-3-(bromomethyl)azetidine, 95%
CAS: 253176-93-1 | C9H16BrNO2 | 250.14 g/mol
Supplier: Thermo Scientific Chemicals H57492MD
Description
It is used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an active pharmaceutical intermediate.
Solubility
Sparingly soluble in water at 25°C, 0.83 g/L.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 1-Boc-3-(bromomethyl)azetidine | |
| C9H16BrNO2 | |
| 250 mg | |
| Sparingly soluble in water at 25°C,0.83g/L. | |
| CC(C)(C)OC(=O)N1CC(CBr)C1 | |
| 250.14 | |
| 250.13 |
| 253176-93-1 | |
| MFCD16556174 | |
| 1-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl azetidine-1-carboxylate, 1-boc-3-bromomethylazetidine, n-tert-butoxycarbonyl-3-bromomethylazetidine, 3-bromomethyl-azetidine-1-carboxylic acid tert-butyl ester, acmc-209d8t, 1-n-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl-1-azetidinecarboxylate, tert-butyl 3-bromomethyl ;azetidine-1-carboxylate, 2-methyl-2-propanyl 3-bromomethyl-1-azetidinecarboxylate | |
| PUKCUGDJEPVLPR-UHFFFAOYSA-N | |
| tert-butyl 3-(bromomethyl)azetidine-1-carboxylate | |
| 53350331 | |
| 95% |
Chemical Identifiers
| 253176-93-1 | |
| 250.14 | |
| PUKCUGDJEPVLPR-UHFFFAOYSA-N | |
| 53350331 | |
| CC(C)(C)OC(=O)N1CC(CBr)C1 |
| C9H16BrNO2 | |
| MFCD16556174 | |
| 1-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl azetidine-1-carboxylate, 1-boc-3-bromomethylazetidine, n-tert-butoxycarbonyl-3-bromomethylazetidine, 3-bromomethyl-azetidine-1-carboxylic acid tert-butyl ester, acmc-209d8t, 1-n-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl-1-azetidinecarboxylate, tert-butyl 3-bromomethyl ;azetidine-1-carboxylate, 2-methyl-2-propanyl 3-bromomethyl-1-azetidinecarboxylate | |
| tert-butyl 3-(bromomethyl)azetidine-1-carboxylate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only