missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Boc-3-aminopyrrolidine, 96%

Catalog No. p-7030506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2923103 1 g
AAH2923106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2923103 Supplier Thermo Scientific Chemicals Supplier No. H2923103
Only null left

CAS: 186550-13-0 | C9H19N2O2 | 187.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 186550-13-0
Molecular Formula C9H19N2O2
Molecular Weight (g/mol) 187.26
MDL Number MFCD01861220
InChI Key CMIBWIAICVBURI-SSDOTTSWSA-O
Synonym 1-boc-3-aminopyrrolidine, 3-amino-1-boc-pyrrolidine, +/--3-amino-1-n-boc-pyrrolidine, 3-amino-1-n-boc-pyrrolidine, 3-amino-1-tert-butoxycarbonyl pyrrolidine, +/--1-boc-3-aminopyrrolidine, 3-amino-pyrrolidine-1-carboxylic acid tert-butyl ester, 1-pyrrolidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, n-boc-3-aminopyrrolidine, 1-bocap
PubChem CID 2756370
IUPAC Name tert-butyl 3-aminopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC[C@@H]([NH3+])C1

Specifications

Density 1.022
Refractive Index 1.471
Quantity 1 g
UN Number UN2735
Sensitivity Air sensitive
Formula Weight 186.25
Percent Purity 96%
Chemical Name or Material (+/-)-1-Boc-3-aminopyrrolidine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only