missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-2-phenylpiperazine, 97%

CAS: 886766-60-5 | C15H22N2O2 | 262.353 g/mol

Supplier:  Thermo Scientific Chemicals H5279703

 View more versions of this product

Catalog No. AAH5279703


Only null left
Explore available promotions

Description

Description

1-Boc-2-phenylpiperazine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-2-phenylpiperazine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

1-Boc-2-phenylpiperazine
C15H22N2O2
1 g
Slightly soluble in water.
CC(C)(C)OC(=O)N1CCNCC1C2=CC=CC=C2
262.353
262.35
886766-60-5
MFCD07367734
1-boc-2-phenylpiperazine, n-1-boc-2-phenyl piperizine, n-1-boc-2-phenylpiperazine, 1-n-boc-2-phenylpiperazine, n-boc-2-bhenyl piperazine, 2-phenylpiperazine, n1-boc protected, 2-phenyl-piperazine-1-carboxylic acid tert-butyl ester, n1-boc-2-phenylpiperazine, n-1-boc-2-phenyl-piperazine, tert-butyl 2-phenylpiperazinecarboxylate
DVOURBIBCQYVCC-UHFFFAOYSA-N
tert-butyl 2-phenylpiperazine-1-carboxylate
16740572
97%

Chemical Identifiers

886766-60-5
262.353
DVOURBIBCQYVCC-UHFFFAOYSA-N
16740572
CC(C)(C)OC(=O)N1CCNCC1C2=CC=CC=C2
C15H22N2O2
MFCD07367734
1-boc-2-phenylpiperazine, n-1-boc-2-phenyl piperizine, n-1-boc-2-phenylpiperazine, 1-n-boc-2-phenylpiperazine, n-boc-2-bhenyl piperazine, 2-phenylpiperazine, n1-boc protected, 2-phenyl-piperazine-1-carboxylic acid tert-butyl ester, n1-boc-2-phenylpiperazine, n-1-boc-2-phenyl-piperazine, tert-butyl 2-phenylpiperazinecarboxylate
tert-butyl 2-phenylpiperazine-1-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only