missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-2-phenylpiperazine, 97%

Catalog No. p-7035091
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52797MD 250 mg
AAH5279703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52797MD Supplier Thermo Scientific Chemicals Supplier No. H52797MD

CAS: 886766-60-5 | C15H22N2O2 | 262.353 g/mol

1-Boc-2-phenylpiperazine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-2-phenylpiperazine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 886766-60-5
Molecular Formula C15H22N2O2
Molecular Weight (g/mol) 262.353
MDL Number MFCD07367734
InChI Key DVOURBIBCQYVCC-UHFFFAOYSA-N
Synonym 1-boc-2-phenylpiperazine, n-1-boc-2-phenyl piperizine, n-1-boc-2-phenylpiperazine, 1-n-boc-2-phenylpiperazine, n-boc-2-bhenyl piperazine, 2-phenylpiperazine, n1-boc protected, 2-phenyl-piperazine-1-carboxylic acid tert-butyl ester, n1-boc-2-phenylpiperazine, n-1-boc-2-phenyl-piperazine, tert-butyl 2-phenylpiperazinecarboxylate
PubChem CID 16740572
IUPAC Name tert-butyl 2-phenylpiperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCNCC1C2=CC=CC=C2

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 262.35
Percent Purity 97%
Chemical Name or Material 1-Boc-2-phenylpiperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only