missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-1,2,3,6-tetrahydropyridine, 97%

Catalog No. p-7034874
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5257403 1 g
AAH5257406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5257403 Supplier Thermo Scientific Chemicals Supplier No. H5257403

CAS: 85838-94-4 | C10H17NO2 | 183.251 g/mol

1-Boc-1,2,3,6-tetrahydropyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-1,2,3,6-tetrahydropyridine is used as pharmaceutical intermediate.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 85838-94-4
Molecular Formula C10H17NO2
Molecular Weight (g/mol) 183.251
MDL Number MFCD04972245
InChI Key SHHHRQFHCPINIB-UHFFFAOYSA-N
Synonym n-boc-1,2,3,6-tetrahydropyridine, tert-butyl 5,6-dihydropyridine-1 2h-carboxylate, tert-butyl 1,2,3,6-tetrahydropyridine-1-carboxylate, tert-butyl 3,6-dihydropyridine-1 2h-carboxylate, 1-boc-1,2,3,6-tetrahydro-pyridine, 1-boc-1,2,3,6-tetrahydropyridine, n-boc-1,2,5,6-tetrahydropyridine, n-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridine, 1 2h-pyridinecarboxylic acid, 3,6-dihydro-, 1,1-dimethylethyl ester, 3,6-dihydro-2h-pyridine-1-carboxylic acid tert-butyl ester
PubChem CID 13094787
IUPAC Name tert-butyl 3,6-dihydro-2H-pyridine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC=CC1

Specifications

Boiling Point 78°C to 84°C (5mmHg)
Solubility Information Insoluble in water. Soluble in organic solvents.
Density 1.029
Quantity 1 g
Formula Weight 183.25
Percent Purity 97%
Chemical Name or Material 1-Boc-1,2,3,6-tetrahydropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only