missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-beta-D-Arabinofuranosyluracil 98.0+%, TCI America™
$29.83 - $50.33
Chemical Identifiers
| CAS | 3083-77-0 |
|---|---|
| Molecular Formula | C9H12N2O6 |
| Molecular Weight (g/mol) | 244.20 |
| MDL Number | MFCD00065998 |
| InChI Key | DRTQHJPVMGBUCF-CCXZUQQUSA-N |
| Synonym | 1-2r,3s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione |
| PubChem CID | 57416896 |
| IUPAC Name | 1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione |
| SMILES | OC[C@H]1O[C@H]([C@@H](O)[C@@H]1O)N1C=CC(=O)NC1=O |
Chemical Identifiers
| 3083-77-0 | |
| 244.20 | |
| DRTQHJPVMGBUCF-CCXZUQQUSA-N | |
| 57416896 | |
| OC[C@H]1O[C@H]([C@@H](O)[C@@H]1O)N1C=CC(=O)NC1=O |
| C9H12N2O6 | |
| MFCD00065998 | |
| 1-2r,3s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione | |
| 1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione |
Specifications
| 3083-77-0 | |
| White-Yellow | |
| MFCD00065998 | |
| 1-2r,3s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione | |
| OC[C@H]1O[C@H]([C@@H](O)[C@@H]1O)N1C=CC(=O)NC1=O | |
| 244.20 | |
| 244.20 | |
| Crystalline Powder |
| 222°C | |
| C9H12N2O6 | |
| 1 g | |
| DRTQHJPVMGBUCF-CCXZUQQUSA-N | |
| 1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
| 57416896 | |
| ≥98.0% (HPLC,T) | |
| 1-beta-D-Arabinofuranosyluracil |
Safety and Handling
RTECSNumber : YQ8818000
TSCA : No
Recommended Storage : Freezer