missing translation for 'onlineSavingsMsg'
Learn More

1-beta-D-Arabinofuranosyluracil 98.0+%, TCI America™

Catalog No. A23561G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23561G 1 g
A23565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23561G Supplier TCI America Supplier No. A23561G

Chemical Identifiers

CAS 3083-77-0
Molecular Formula C9H12N2O6
Molecular Weight (g/mol) 244.20
MDL Number MFCD00065998
InChI Key DRTQHJPVMGBUCF-CCXZUQQUSA-N
Synonym 1-2r,3s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione
PubChem CID 57416896
IUPAC Name 1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H]([C@@H](O)[C@@H]1O)N1C=CC(=O)NC1=O

Specifications

Melting Point 222°C
Color White-Yellow
Quantity 1 g
Formula Weight 244.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-beta-D-Arabinofuranosyluracil
RTECSNumber YQ8818000
TSCA No
Recommended Storage Freezer