missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-beta-D-Arabinofuranosyluracil, 99%
CAS: 3083-77-0 | C9H12N2O6 | 244.20 g/mol
$33.90 - $275.38
Chemical Identifiers
| CAS | 3083-77-0 |
|---|---|
| Molecular Formula | C9H12N2O6 |
| Molecular Weight (g/mol) | 244.20 |
| MDL Number | MFCD00065998 |
| InChI Key | DRTQHJPVMGBUCF-CCXZUQQUSA-N |
| Synonym | 1-2r,3s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione |
| PubChem CID | 57416896 |
| SMILES | OC[C@H]1O[C@H]([C@@H](O)[C@@H]1O)N1C=CC(=O)NC1=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6414703
|
Thermo Scientific Chemicals
J6414703 |
1 g |
Each for $33.90
|
|
|||||
|
AAJ6414706
|
Thermo Scientific Chemicals
J6414706 |
5 g |
Each for $87.90
|
|
|||||
|
AAJ6414714
|
Thermo Scientific Chemicals
J6414714 |
25 g |
Each for $275.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3083-77-0 | |
| 244.20 | |
| DRTQHJPVMGBUCF-CCXZUQQUSA-N | |
| 57416896 |
| C9H12N2O6 | |
| MFCD00065998 | |
| 1-2r,3s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione | |
| OC[C@H]1O[C@H]([C@@H](O)[C@@H]1O)N1C=CC(=O)NC1=O |
Specifications
| 3083-77-0 | |
| C9H12N2O6 | |
| 1 g | |
| 1-2r,3s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione | |
| OC[C@H]1O[C@H]([C@@H](O)[C@@H]1O)N1C=CC(=O)NC1=O | |
| 244.20 | |
| 244.2 | |
| Powder |
| White | |
| MFCD00065998 | |
| 28749 | |
| DRTQHJPVMGBUCF-CCXZUQQUSA-N | |
| 1-[(2R,3S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione | |
| 57416896 | |
| 99% | |
| 1-beta-D-Arabinofuranosyluracil |
Safety and Handling
EINECSNumber : 221-386-9
RTECSNumber : YQ8818000
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only