missing translation for 'onlineSavingsMsg'
Learn More

1-[(Benzyloxy)carbonyl]-2-indolinecarboxylic acid, ≥90%, Thermo Scientific™

Chemical Identifiers

CAS 117483-89-3
Molecular Formula C17H15NO4
Molecular Weight (g/mol) 297.31
MDL Number MFCD02682032
InChI Key BSOYWTITVKXHLM-UHFFFAOYNA-N
Synonym 1-benzyloxy carbonyl-2-indolinecarboxylic acid, 1-benzyloxy carbonyl indoline-2-carboxylic acid, 1-benzyloxy carbonyl-2,3-dihydroindole-2-carboxylic acid, 2,3-dihydro-1h-indole-1,2-dicarboxylic acid 1-benzyl ester, 1-benzyloxy carbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, acmc-1brpx, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, 2s, 1-benzyloxycarbonyl-2-indoline carboxylic acid, 1-benzyl-oxy carbonyl-2-indolinecarboxylic acid
PubChem CID 2776400
IUPAC Name 1-phenylmethoxycarbonyl-2,3-dihydroindole-2-carboxylic acid
SMILES OC(=O)C1CC2=CC=CC=C2N1C(=O)OCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC13401DA
SDS View Documents
Thermo Scientific Chemicals
CC13401DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC13401DE
SDS View Documents
Thermo Scientific Chemicals
CC13401DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

117483-89-3
297.31
BSOYWTITVKXHLM-UHFFFAOYNA-N
2776400
OC(=O)C1CC2=CC=CC=C2N1C(=O)OCC1=CC=CC=C1
C17H15NO4
MFCD02682032
1-benzyloxy carbonyl-2-indolinecarboxylic acid, 1-benzyloxy carbonyl indoline-2-carboxylic acid, 1-benzyloxy carbonyl-2,3-dihydroindole-2-carboxylic acid, 2,3-dihydro-1h-indole-1,2-dicarboxylic acid 1-benzyl ester, 1-benzyloxy carbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, acmc-1brpx, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, 2s, 1-benzyloxycarbonyl-2-indoline carboxylic acid, 1-benzyl-oxy carbonyl-2-indolinecarboxylic acid
1-phenylmethoxycarbonyl-2,3-dihydroindole-2-carboxylic acid

Specifications

117483-89-3
100.0
Amber glass bottle
MFCD02682032
1-benzyloxy carbonyl-2-indolinecarboxylic acid, 1-benzyloxy carbonyl indoline-2-carboxylic acid, 1-benzyloxy carbonyl-2,3-dihydroindole-2-carboxylic acid, 2,3-dihydro-1h-indole-1,2-dicarboxylic acid 1-benzyl ester, 1-benzyloxy carbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, acmc-1brpx, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, 2s, 1-benzyloxycarbonyl-2-indoline carboxylic acid, 1-benzyl-oxy carbonyl-2-indolinecarboxylic acid
OC(=O)C1CC2=CC=CC=C2N1C(=O)OCC1=CC=CC=C1
297.31
297.31
1-[(Benzyloxy)carbonyl]-2-indolinecarboxylic acid
89.5
90%+
C17H15NO4
1g
BSOYWTITVKXHLM-UHFFFAOYNA-N
1-phenylmethoxycarbonyl-2,3-dihydroindole-2-carboxylic acid
2776400
90%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'