missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyloxy-4-nitrobenzene 98.0+%, TCI America™

Catalog No. B47065G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B47065G Supplier TCI America Supplier No. B47065G

Chemical Identifiers

CAS 1145-76-2
Molecular Formula C13H11NO3
Molecular Weight (g/mol) 229.24
MDL Number MFCD00024672
InChI Key YOVUXLHIVNBVKO-UHFFFAOYSA-N
Synonym Benzyl 4-Nitrophenyl Ether
PubChem CID 70842
IUPAC Name 1-(benzyloxy)-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 5 g
Formula Weight 229.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyloxy-4-nitrobenzene