missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyloxy-4-bromobenzene 98.0+%, TCI America™
$46.20 - $198.30
Chemical Identifiers
| CAS | 6793-92-6 |
|---|---|
| Molecular Formula | C13H11BrO |
| Molecular Weight (g/mol) | 263.134 |
| MDL Number | MFCD00028016 |
| InChI Key | OUQSGILAXUXMGI-UHFFFAOYSA-N |
| Synonym | 4-benzyloxybromobenzene, 1-benzyloxy-4-bromobenzene, 1-bromo-4-phenylmethoxy benzene, 1-bromo-4-benzyloxybenzene, 4-benzyloxy bromobenzene, 1-benzyloxy-4-bromo-benzene, 1-bromo-4-phenylmethyl oxy benzene, 4-benzyloxy-bromobenzene, p-benzyloxy bromobenzene |
| PubChem CID | 138835 |
| IUPAC Name | 1-bromo-4-phenylmethoxybenzene |
| SMILES | C1=CC=C(C=C1)COC2=CC=C(C=C2)Br |
Chemical Identifiers
| 6793-92-6 | |
| 263.134 | |
| OUQSGILAXUXMGI-UHFFFAOYSA-N | |
| 138835 | |
| C1=CC=C(C=C1)COC2=CC=C(C=C2)Br |
| C13H11BrO | |
| MFCD00028016 | |
| 4-benzyloxybromobenzene, 1-benzyloxy-4-bromobenzene, 1-bromo-4-phenylmethoxy benzene, 1-bromo-4-benzyloxybenzene, 4-benzyloxy bromobenzene, 1-benzyloxy-4-bromo-benzene, 1-bromo-4-phenylmethyl oxy benzene, 4-benzyloxy-bromobenzene, p-benzyloxy bromobenzene | |
| 1-bromo-4-phenylmethoxybenzene |
Specifications
| 6793-92-6 | |
| White-Yellow | |
| MFCD00028016 | |
| 4-benzyloxybromobenzene, 1-benzyloxy-4-bromobenzene, 1-bromo-4-phenylmethoxy benzene, 1-bromo-4-benzyloxybenzene, 4-benzyloxy bromobenzene, 1-benzyloxy-4-bromo-benzene, 1-bromo-4-phenylmethyl oxy benzene, 4-benzyloxy-bromobenzene, p-benzyloxy bromobenzene | |
| C1=CC=C(C=C1)COC2=CC=C(C=C2)Br | |
| 263.134 | |
| 263.13 | |
| Crystalline Powder |
| 62°C | |
| C13H11BrO | |
| 5 g | |
| OUQSGILAXUXMGI-UHFFFAOYSA-N | |
| 1-bromo-4-phenylmethoxybenzene | |
| 138835 | |
| ≥98.0% (GC) | |
| 1-Benzyloxy-4-bromobenzene |
Safety and Handling
TSCA : No