missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyloxy-4-bromobenzene 98.0+%, TCI America™

Catalog No. B21275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21275G 5 g
B212725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21275G Supplier TCI America Supplier No. B21275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6793-92-6
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.134
MDL Number MFCD00028016
InChI Key OUQSGILAXUXMGI-UHFFFAOYSA-N
Synonym 4-benzyloxybromobenzene, 1-benzyloxy-4-bromobenzene, 1-bromo-4-phenylmethoxy benzene, 1-bromo-4-benzyloxybenzene, 4-benzyloxy bromobenzene, 1-benzyloxy-4-bromo-benzene, 1-bromo-4-phenylmethyl oxy benzene, 4-benzyloxy-bromobenzene, p-benzyloxy bromobenzene
PubChem CID 138835
IUPAC Name 1-bromo-4-phenylmethoxybenzene
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)Br

Specifications

Melting Point 62°C
Color White-Yellow
Quantity 5 g
Formula Weight 263.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyloxy-4-bromobenzene