missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyloxy-3-iodobenzene 97.0+%, TCI America™

Catalog No. B46895G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B46895G Supplier TCI America Supplier No. B46895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107623-21-2
Molecular Formula C13H11IO
Molecular Weight (g/mol) 310.13
MDL Number MFCD01318100
InChI Key QMKHOPJXDQAHBG-UHFFFAOYSA-N
Synonym 1-benzyloxy-3-iodobenzene, 3-iodobenzyloxybenzene, 1-benzyloxy-3-iodo-benzene, benzene,1-iodo-3-phenylmethoxy, benzene, 1-iodo-3-phenylmethoxy, pubchem3066, 3-benzyloxyiodobenzene, acmc-1c7zz, 1-benzyl-oxy-3-iodobenzene
PubChem CID 4228019
IUPAC Name 1-(benzyloxy)-3-iodobenzene
SMILES IC1=CC=CC(OCC2=CC=CC=C2)=C1

Specifications

Melting Point 54°C
Quantity 5 g
Formula Weight 310.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyloxy-3-iodobenzene