missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyloxy-3-iodobenzene, 99%

Catalog No. AAL1744206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1744206 5 g
AAL1744203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1744206 Supplier Thermo Scientific Chemicals Supplier No. L1744206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107623-21-2 | C13H11IO | 310.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107623-21-2
Molecular Formula C13H11IO
Molecular Weight (g/mol) 310.13
MDL Number MFCD01318100
InChI Key QMKHOPJXDQAHBG-UHFFFAOYSA-N
Synonym 1-benzyloxy-3-iodobenzene, 3-iodobenzyloxybenzene, 1-benzyloxy-3-iodo-benzene, benzene,1-iodo-3-phenylmethoxy, benzene, 1-iodo-3-phenylmethoxy, pubchem3066, 3-benzyloxyiodobenzene, acmc-1c7zz, 1-benzyl-oxy-3-iodobenzene
PubChem CID 4228019
IUPAC Name 1-iodo-3-phenylmethoxybenzene
SMILES IC1=CC=CC(OCC2=CC=CC=C2)=C1

Specifications

Melting Point 50°C to 51°C
Quantity 5 g
Beilstein 3270112
Sensitivity Light sensitive
Formula Weight 310.14
Percent Purity 99%
Chemical Name or Material 1-Benzyloxy-3-iodobenzene
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only