missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyloxy-3-bromobenzene 98.0+%, TCI America™
$63.38 - $249.78
Chemical Identifiers
| CAS | 53087-13-1 |
|---|---|
| Molecular Formula | C13H11BrO |
| Molecular Weight (g/mol) | 263.134 |
| MDL Number | MFCD00155065 |
| InChI Key | HVWZMGZBJCJDOX-UHFFFAOYSA-N |
| Synonym | 1-benzyloxy-3-bromobenzene, 3-benzyloxybromobenzene, benzyl 3-bromophenyl ether, 1-bromo-3-benzyloxybenzene, 3-benzyloxy bromobenzene, 3-bromophenyl benzyl ether, benzene, 1-bromo-3-phenylmethoxy, 1-benzyloxy-3-bromo-benzene, pubchem3065 |
| PubChem CID | 2756638 |
| IUPAC Name | 1-bromo-3-phenylmethoxybenzene |
| SMILES | C1=CC=C(C=C1)COC2=CC(=CC=C2)Br |
Chemical Identifiers
| 53087-13-1 | |
| 263.134 | |
| HVWZMGZBJCJDOX-UHFFFAOYSA-N | |
| 2756638 | |
| C1=CC=C(C=C1)COC2=CC(=CC=C2)Br |
| C13H11BrO | |
| MFCD00155065 | |
| 1-benzyloxy-3-bromobenzene, 3-benzyloxybromobenzene, benzyl 3-bromophenyl ether, 1-bromo-3-benzyloxybenzene, 3-benzyloxy bromobenzene, 3-bromophenyl benzyl ether, benzene, 1-bromo-3-phenylmethoxy, 1-benzyloxy-3-bromo-benzene, pubchem3065 | |
| 1-bromo-3-phenylmethoxybenzene |
Specifications
| 53087-13-1 | |
| White | |
| MFCD00155065 | |
| 1-benzyloxy-3-bromobenzene, 3-benzyloxybromobenzene, benzyl 3-bromophenyl ether, 1-bromo-3-benzyloxybenzene, 3-benzyloxy bromobenzene, 3-bromophenyl benzyl ether, benzene, 1-bromo-3-phenylmethoxy, 1-benzyloxy-3-bromo-benzene, pubchem3065 | |
| C1=CC=C(C=C1)COC2=CC(=CC=C2)Br | |
| 263.134 | |
| 263.13 | |
| Crystalline Powder |
| 62°C | |
| C13H11BrO | |
| 5 g | |
| HVWZMGZBJCJDOX-UHFFFAOYSA-N | |
| 1-bromo-3-phenylmethoxybenzene | |
| 2756638 | |
| ≥98.0% (GC) | |
| 1-Benzyloxy-3-bromobenzene |
Safety and Handling
TSCA : No